Tisk…

Anti-RAB3D Rabbit Monoclonal Antibody [clone: EPR8107]

Dodavatel: Abcam
ABCAAB133301-100EA 23840 CZK
ABCAAB133301-100
Anti-RAB3D Rabbit Monoclonal Antibody [clone: EPR8107]
Protilátky
Type: Primary
Antigen: RAB3D (RAB3D, member RAS oncogene family)
Clonality: monoclonal
Clone: EPR8107
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR