Tisk…

Anti-YES1 Rabbit Monoclonal Antibody [clone: EPR3172]

Dodavatel: Abcam
ABCAAB133314-100EA 25620 CZK
ABCAAB133314-100
Anti-YES1 Rabbit Monoclonal Antibody [clone: EPR3172]
Protilátky
Type: Primary
Antigen: YES1 (v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1)
Clonality: monoclonal
Clone: EPR3172
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR