Tisk…

Anti-ARPC2 Rabbit Monoclonal Antibody [clone: EPR8533]

Dodavatel: Abcam
ABCAAB133315-100EA 23960 CZK
ABCAAB133315-100
Anti-ARPC2 Rabbit Monoclonal Antibody [clone: EPR8533]
Protilátky
Type: Primary
Antigen: ARPC2 (actin related protein 2/3 complex, subunit 2, 34kDa)
Clonality: monoclonal
Clone: EPR8533
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR