Tisk…

Anti-ITGB4 Rabbit Monoclonal Antibody [clone: EPR6513]

Dodavatel: Abcam
ABCAAB133321-100EA 24060 CZK
ABCAAB133321-100
Anti-ITGB4 Rabbit Monoclonal Antibody [clone: EPR6513]
Protilátky
Type: Primary
Antigen: ITGB4 (integrin, beta 4)
Clonality: monoclonal
Clone: EPR6513
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR