Tisk…
Anti-SLC7A2 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB133368-50EA 24390 CZK
ABCAAB133368-50
Anti-SLC7A2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC7A2 (solute carrier family 7 (cationic amino acid transporter, y+ system), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human