Tisk…

Anti-SURF4 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB133369-50EA 25740 CZK
ABCAAB133369-50
Anti-SURF4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SURF4 (surfeit 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR