Tisk…
Anti-LRRN4 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB133372-50EA 24420 CZK
ABCAAB133372-50
Anti-LRRN4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LRRN4 (leucine rich repeat neuronal 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human