Tisk…
Anti-GAB1 Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB133486-100EA 26170 CZK
ABCAAB133486-100
Anti-GAB1 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: GAB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat