Tisk…

Anti-IL22 Rabbit Monoclonal Antibody

Dodavatel: Abcam
ABCAAB133545-100EA 25620 CZK
ABCAAB133545-100
Anti-IL22 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: IL22
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR