Tisk…

Anti-NTNG2 Rabbit Monoclonal Antibody [clone: EPR4570]

Dodavatel: Abcam
ABCAAB133732-100EA 23900 CZK
ABCAAB133732-100
Anti-NTNG2 Rabbit Monoclonal Antibody [clone: EPR4570]
Protilátky
Type: Primary
Antigen: NTNG2 (netrin G2)
Clonality: monoclonal
Clone: EPR4570
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR