Tisk…

Anti-TAF15 Rabbit Monoclonal Antibody [clone: EPR9196]

Dodavatel: Abcam
ABCAAB133760-100EA 26170 CZK
ABCAAB133760-100
Anti-TAF15 Rabbit Monoclonal Antibody [clone: EPR9196]
Protilátky
Type: Primary
Antigen: TAF15 (TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa)
Clonality: monoclonal
Clone: EPR9196
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR