Tisk…

Anti-IL22 Mouse Monoclonal Antibody [clone: IMG8F11E2]

Dodavatel: Abcam
ABCAAB134035-100EA 26320 CZK
ABCAAB134035-100
Anti-IL22 Mouse Monoclonal Antibody [clone: IMG8F11E2]
Protilátky
Type: Primary
Antigen: IL22 (interleukin 22)
Clonality: monoclonal
Clone: IMG8F11E2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR