Tisk…

Anti-GHR Rabbit monoclonal antibody unconjugated [clone: EPR5291]

Dodavatel: Abcam
ABCAAB134078-100EA 26350 CZK
ABCAAB134078-100
Anti-GHR Rabbit monoclonal antibody unconjugated [clone: EPR5291]
Protilátky
Type: Primary
Antigen: GHR (growth hormone receptor)
Clonality: monoclonal
Clone: EPR5291
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR