Tisk…
Anti-Mannose 6 Phosphate Receptor (Cation dependent) Rabbit Monoclonal Antibody [clone: EPR7691]
Dodavatel: Abcam
ABCAAB134153-100EA 28710 CZK
ABCAAB134153-100
Anti-Mannose 6 Phosphate Receptor (Cation dependent) Rabbit Monoclonal Antibody [clone: EPR7691]
Protilátky
Type: Primary
Antigen: Mannose 6 Phosphate Receptor (Cation dependent)
Clonality: monoclonal
Clone: EPR7691
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat