Tisk…
Anti-NCL Rabbit Monoclonal Antibody [clone: EPR7951]
Dodavatel: Abcam
ABCAAB134164-100EA 27150 CZK
ABCAAB134164-100
Anti-NCL Rabbit Monoclonal Antibody [clone: EPR7951]
Protilátky
Type: Primary
Antigen: NCL (nucleolin)
Clonality: monoclonal
Clone: EPR7951
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat