Tisk…

Anti-GSN Rabbit Monoclonal Antibody [clone: EP1940Y]

Dodavatel: Abcam
ABCAAB134183-100EA 26720 CZK
ABCAAB134183-100
Anti-GSN Rabbit Monoclonal Antibody [clone: EP1940Y]
Protilátky
Type: Primary
Antigen: GSN (gelsolin)
Clonality: monoclonal
Clone: EP1940Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR