Tisk…
Anti-ETFA Mouse Monoclonal Antibody [clone: GP1.4]
Dodavatel: Abcam
ABCAAB134282-50EA 26140 CZK
ABCAAB134282-50
Anti-ETFA Mouse Monoclonal Antibody [clone: GP1.4]
Protilátky
Type: Primary
Antigen: ETFA (electron-transfer-flavoprotein, alpha polypeptide)
Clonality: monoclonal
Clone: GP1.4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human