Tisk…

Anti-CD68 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: Y1/82A]

Dodavatel: Abcam
ABCAAB134351-50EA 12960 CZK
ABCAAB134351-50
Anti-CD68 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: Y1/82A]
Protilátky
Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: monoclonal
Clone: Y1/82A
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR