Tisk…

Anti-CD40 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5C3]

Dodavatel: Abcam
ABCAAB134380-50EA 25560 CZK
ABCAAB134380-50
Anti-CD40 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5C3]
Protilátky
Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: monoclonal
Clone: 5C3
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR