Tisk…

Anti-NEFL Chicken Polyclonal Antibody

Dodavatel: Abcam
ABCAAB134460-100EA 26320 CZK
ABCAAB134460-100
Anti-NEFL Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: NEFL (neurofilament, light polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR