Tisk…

Anti-IL2RA Mouse Monoclonal Antibody PE (Phycoerythrin) [clone: BC96]

Dodavatel: Abcam
ABCAAB134476-50EA 23350 CZK
ABCAAB134476-50
Anti-IL2RA Mouse Monoclonal Antibody PE (Phycoerythrin) [clone: BC96]
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: BC96
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR