Tisk…

Anti-POLR3E Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB134560-100EA 24420 CZK
ABCAAB134560-100
Anti-POLR3E Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: POLR3E (polymerase (RNA) III (DNA directed) polypeptide E (80kD))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR