Tisk…

Anti-MAPK9 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB134567-200EA 25740 CZK
ABCAAB134567-200
Anti-MAPK9 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR