Tisk…

Anti-TBC1D4 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB134749-100EA 25740 CZK
ABCAAB134749-100
Anti-TBC1D4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TBC1D4 (TBC1 domain family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR