Tisk…
Anti-LYSYL TRNA SYNTHE Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB134912-100EA 24120 CZK
ABCAAB134912-100
Anti-LYSYL TRNA SYNTHE Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: LYSYL TRNA SYNTHE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat