Tisk…
Anti-C1 inactivator Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB134918-100EA 25950 CZK
ABCAAB134918-100
Anti-C1 inactivator Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: C1 inactivator
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat