Tisk…

Anti-S-Adenosyl-L-homocysteine Rabbit Monoclonal Antibody

Dodavatel: Abcam
ABCAAB134966-100EA 26050 CZK
ABCAAB134966-100
Anti-S-Adenosyl-L-homocysteine Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: S-Adenosyl-L-homocysteine
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR