Tisk…

Anti-ZNF259 Rabbit Monoclonal Antibody

Dodavatel: Abcam
ABCAAB134970-100EA 24060 CZK
ABCAAB134970-100
Anti-ZNF259 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ZNF259
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR