Tisk…
Anti-C5a/C5a Mouse Monoclonal Antibody [clone: MM0608-6H13]
Dodavatel: Abcam
ABCAAB135187-100EA 25590 CZK
ABCAAB135187-100
Anti-C5a/C5a Mouse Monoclonal Antibody [clone: MM0608-6H13]
Protilátky
Type: Primary
Antigen: C5a/C5a
Clonality: monoclonal
Clone: MM0608-6H13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human