Tisk…

Anti-SHH Mouse Monoclonal Antibody [clone: 5H4]

Dodavatel: Abcam
ABCAAB135240-100EA 26630 CZK
ABCAAB135240-100
Anti-SHH Mouse Monoclonal Antibody [clone: 5H4]
Protilátky
Type: Primary
Antigen: SHH (sonic hedgehog)
Clonality: monoclonal
Clone: 5H4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR