Tisk…

Anti-IgG Rabbit Polyclonal Antibody (bGal (beta Galactosidase))

Dodavatel: Abcam
ABCAAB136712-500EA 13730 CZK
ABCAAB136712-500
Anti-IgG Rabbit Polyclonal Antibody (bGal (beta Galactosidase))
Protilátky
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: beta-Galactosidase
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR