Tisk…

Anti-GPR17 Goat Polyclonal Antibody

Dodavatel: Abcam
ABCAAB136902-50EA 25740 CZK
ABCAAB136902-50
Anti-GPR17 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GPR17 (G protein-coupled receptor 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: none
Reactivity: Bovine, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR