Tisk…

Anti-NOS2 Rabbit Monoclonal Antibody [clone: K13-A]

Dodavatel: Abcam
ABCAAB136918-250EA 27030 CZK
ABCAAB136918-250
Anti-NOS2 Rabbit Monoclonal Antibody [clone: K13-A]
Protilátky
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: monoclonal
Clone: K13-A
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR