Tisk…
Anti-PEX19 Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB137072-100EA 27060 CZK
ABCAAB137072-100
Anti-PEX19 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PEX19
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat