Tisk…
Anti-RNF128 Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB137088-100EA 26170 CZK
ABCAAB137088-100
Anti-RNF128 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: RNF128
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat