Tisk…
Anti-PLSCR3 Rabbit Monoclonal Antibody
Dodavatel: Abcam
ABCAAB137128-100EA 25160 CZK
ABCAAB137128-100
Anti-PLSCR3 Rabbit Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PLSCR3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat