Tisk…

Anti-SLC6A8 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137198-100EA 25740 CZK
ABCAAB137198-100
Anti-SLC6A8 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SLC6A8 (solute carrier family 6 (neurotransmitter transporter), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR