Tisk…
Anti-CASR Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB137408-100EA 26540 CZK
ABCAAB137408-100
Anti-CASR Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CASR (calcium-sensing receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human