Tisk…

Anti-FBL Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137422-50EA 26320 CZK
ABCAAB137422-50
Anti-FBL Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FBL (fibrillarin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR