Tisk…

Anti-CYLD Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137524-50EA 26480 CZK
ABCAAB137524-50
Anti-CYLD Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CYLD (cylindromatosis (turban tumor syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR