Tisk…

Anti-LOR Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137533-50EA 25740 CZK
ABCAAB137533-50
Anti-LOR Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LOR (loricrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR