Tisk…

Anti-SFRP2 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137560-100EA 26320 CZK
ABCAAB137560-100
Anti-SFRP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SFRP2 (secreted frizzled-related protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR