Tisk…

Anti-CREB3L1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137565-100EA 25740 CZK
ABCAAB137565-100
Anti-CREB3L1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CREB3L1 (cAMP responsive element binding protein 3-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR