Tisk…

Anti-PPT1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137578-50EA 25740 CZK
ABCAAB137578-50
Anti-PPT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PPT1 (palmitoyl-protein thioesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR