Tisk…

Anti-MAPK15 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137619-100EA 26320 CZK
ABCAAB137619-100
Anti-MAPK15 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAPK15 (mitogen-activated protein kinase 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR