Tisk…
Anti-GNB1 Rabbit Polyclonal Antibody
Dodavatel: Abcam
ABCAAB137635-50EA 26450 CZK
ABCAAB137635-50
Anti-GNB1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GNB1 (guanine nucleotide binding protein (G protein), beta polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse