Tisk…

Anti-PFKP Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137636-50EA 25740 CZK
ABCAAB137636-50
Anti-PFKP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PFKP (phosphofructokinase, platelet)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR