Tisk…

Anti-SCYL1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137639-100EA 24420 CZK
ABCAAB137639-100
Anti-SCYL1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SCYL1 (SCY1-like 1 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR