Tisk…

Anti-ADRBK1 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137666-100EA 26410 CZK
ABCAAB137666-100
Anti-ADRBK1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ADRBK1 (adrenergic, beta, receptor kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR