Tisk…

Anti-HDAC3 Rabbit Polyclonal Antibody

Dodavatel: Abcam
ABCAAB137704-100EA 26320 CZK
ABCAAB137704-100
Anti-HDAC3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR